Tables/workbenches with multi-bar under shelf for optimum airflow
Supplier: TEKNOMEK
Certificates
About this item
This stainless steel workbench features a multi-bar under shelf which helps maintain cleanroom airflow and is perfect for storing items out of the way, but close to hand for when they are needed.
- Fully welded 304-grade stainless steel, brushed dull polish finish
- Supplied fully welded, no assembly
- Multi-bar under shelf for optimum airflow
- Choose from static or mobile with BZP or stainless steel castors
- Added strength
304 grade stainless steel, sturdy 40 mm box section frame, designed with hygiene in mind.
Load capacity: 225 kg (for models with castors) and 250 kg (for models with feet).
Frequently Bought Together