Fluoribacter bozemanae CECT® 7276 Vitroids™, Certified Reference Materials, Suitable for Microbiology, Supelco®
Catalog # VT072767-10EA
Supplier: Merck
CAS Number:
Specifications
- Pk:10
- Description:Fluoribacter bozemanae CECT® 7276 Vitroids™, >100000 CFU mean value range
- M-Clarity quality level:MQ300
- Storage temperature:–20 °C
- Product type:Hydrosoluble discs
Specifications
About this item
Derived from CECT® strains.
Certifications: These certified reference materials (CRMs) are produced and certified in accordance with ISO 17034 and ISO/IEC 17025.