Order Entry
Czech Republic
ContactUsLinkComponent
Single Element Standard for AA, Iron
Single Element Standard for AA, Iron
Catalog # HEWL5190-8285
Supplier:  Agilent
CAS Number:  
undefined
Single Element Standard for AA, Iron
Catalog # HEWL5190-8285
Supplier:  Agilent
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    5% HNO₃
  • Pk:
    100 mL
  • Element:
    Iron
  • Symbol:
    Fe

Specifications

About this item

To ensure the stability and performance of atomic absorption analyses, Agilent offers a comprehensive portfolio of single-element and multi-element reference materials that test the efficacy of the instrument.

  • Gives you confidence in your AAS
  • Accurate, reliable calibrations

Agilent calibration standards are of the highest quality and are designed, developed, and manufactured to guarantee at least one year of shelf life.

Delivery: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.