About this item
To avoid spill over of canisters through the connection of a b.safe Control Waste Cap to a b.safe Display for electronic level control.
- PPS screw cap
- Freely rotatable insert made of PTFE
- Avoid overflowing canisters
With 2 permanently fixed cables (length approx. 0,5 m each) for the connection to a b.safe Display which signals a pre-alert when approx. 50% of the nominal volume are reached and a main alert for canister change as soon as 100% of the nominal volume are reached. With 1 additional red LED flashing light for easy identification of a full canister. With PFA fittings for capillary tubes with OD 1,6/2,2/3,2 mm (thread UNF 1/4“), hose connector for tubing ID 5,5-8 mm (thread NPT 1/8“), blind fittings for closing unused connections and 1 connector GL 14 for fixing the b.safe Exhaust Filter.
The additional pre-alert enables the user to provide exchange containers for big volumes of waste e.g. before the end of work or the weekend. Reliable monitoring of the filling levels of waste canisters in laboratory cupboards and workbenches. With the use of b.safe Waste Tubes, solvent waste is led directly into the canister, solvent vapours are held back by the exhaust filter with activated carbon or can be led to the hood by means of a venting hose.