ContactUsLinkComponent
System waste caps capacitive, b.safe
  BOHLR1325-60
 :  Bohlender
System waste caps capacitive, b.safe
  BOHLR1325-60
 :  Bohlender
  • Thread:
    S 60
  • Description:
    Waste caps
  • Material:
    PPS
  • Connector:
    1× GL 25 (m) hose connector, 1× GL 15 (f) aeration connector
  • Pk:
    1
To avoid spill over of canisters through the connection of a waste cap capacitive to a b.safe Display for electronic level control. With the use of b.safe waste tubes, solvent waste is led directly into the canister, solvent vapours are held back by the exhaust filter with activated carbon or can be led to the hood by means of a venting hose.
  • Made of high-performance plastics
  • Thermally and chemically stable
  • Good grip for easy assembly, safe working and improved processes
Screw cap made of PPS, free movable insert made of conductive PE-EX, permanently fixed sensor for capacitive level control with cable (0,5 m). With 1 hose connector GL 25 suitable for b.safe Waste Tubes and 1 connector GL14 for fixing the b.safe Venting Hose or a b.safe Exhaust Filter.
Design and scope of delivery according to table.
Exhaust filters