Transparent Cryogenic Tapes for Frozen Surfaces
: GA
Transparent tape for labelling already frozen containers under deep freeze conditions (–80 °C). This cryogenic tape can be inscribed with permanent cryogenic markers, and is ideal for identifying containers intended for long-term storage in cryogenic (–196 °C) and deep freeze (–80 °C) conditions.
- Temperature range: –196 to +150 °C (–321 to +302 °F)
- Tape can adhere to already frozen containers and boxes
- Ideal for long-term storage in cryogenic (–196 °C) and deep freeze (–80 °C) conditions
- Strong adhesion to surfaces, even in water bath thawing applications
- Can easily be inscribed with permanent cryo markers
This transparent cryogenic tape for frozen surfaces comes in a roll format to identify already frozen plastic containers. The tape is designed for long-term cryo-preservation in liquid nitrogen (–196 °C), ultra-low temperature freezers (–80 °C), and dry ice. These tapes are waterproof and suitable for immersion in a water bath. Can be conveniently inscribed with permanent cryogenic markers.
Frequently Bought Together