ESD safety shoes, lace-up, Larkin XXSports Low
Supplier: ELTEN
Certificates
About this item
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
- Padded tongue
- TPU/PU sole WELLMAXX SPORTS
- Steel toe cap
- Leather-free
Mesh material construction with fabric lining for natural ventilation. Full-length inlay sole, SPORTIVE ESD. Non-metallic, penetration-resistant midsole. Sole core made of Infinergy® by BASF.
Certifications: EN ISO 20345 S1P SRA, form A.
Specifications
- Catalog No:
- ELTN728101/38
- ELTN728101/36
- ELTN728101/37
- ELTN728101/35
- ELTN728111/35
- ELTN728111/36
- ELTN728101/47
- ELTN728101/48
- ELTN728101/41
- ELTN728111/37
- ELTN728111/47
- ELTN728101/42
- ELTN728111/48
- ELTN728111/38
- ELTN728111/39
- ELTN728101/39
- ELTN728101/40
- ELTN728111/40
- ELTN728111/41
- ELTN728101/45
- ELTN728101/46
- ELTN728111/42
- ELTN728101/43
- ELTN728111/43
- ELTN728101/44
- ELTN728111/44
- ELTN728111/45
- ELTN728111/46
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001