ESD safety shoes, lace-up, S1P, Marten XXSports Low
: ELTEN
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
- Full, padded bellows tongue
- TPU/PU sole WELLMAXX SPORTS
- Steel toe cap
- Extra laces included
Microfibre/mesh material construction with fabric lining for natural ventilation. Full-length inlay sole, SPORTIVE ESD. Non-metallic, penetration-resistant midsole. Lacing system with individual quick locking system. Sole core made of Infinergy® by BASF.
: EN ISO 20345 S1P SRA, form A.
- Catalog No:
- ELTN728121/44
- ELTN728121/38
- ELTN728121/39
- ELTN728121/36
- ELTN728121/47
- ELTN728121/37
- ELTN728121/48
- ELTN728121/42
- ELTN728121/43
- ELTN728121/40
- ELTN728121/41
- ELTN728121/45
- ELTN728121/46
- ELTN728121/35
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
Frequently Bought Together