ESD safety shoes, BOA® Fit System, HANNAH XXTL BOA® Low
Supplier: ELTEN
Certificates
About this item
Ladies' safety shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
- Full, padded bellows tongue
- PU/PU sole, WELLMAXX TRAINERS LADY
- Steel toe cap
- BOA® Fit System
- Leather-free
Hydrophobised microfibre/hydrophobised textile material construction with fabric lining for natural ventilation.Full-length inlay sole, ESD PRO Lady Y.Non-metallic, penetration-resistant midsole.Sole core made of Infinergy® by BASF.
Certifications: EN ISO 20345 S3 SRC, form A.
Specifications
- Catalog No:
- ELTN742561/35
- ELTN742561/36
- ELTN742561/39
- ELTN742561/40
- ELTN742561/37
- ELTN742561/38
- ELTN742561/41
- ELTN742561/42
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001