ContactUsLinkComponent
Human cord blood endothelial progenitor cell (hCB-EPC), CELLvo™
Human cord blood endothelial progenitor cell (hCB-EPC), CELLvo™
Supplier:  STEMBIOSYS
Certificates
undefined
Human cord blood endothelial progenitor cell (hCB-EPC), CELLvo™
Supplier:  STEMBIOSYS
Specifications
  • Storage conditions:
    Upon arrival, store cells in liquid nitrogen. CELLvo™ Matrix should be stored at 2...8 °C upon arrival. Avoid extended exposure to light.
  • Storage buffer:
    CryoStor®CS5