Order Entry
Czech Republic
ContactUsLinkComponent
Matrix Plus, CELLvo™
Matrix Plus, CELLvo™
Supplier:  STEMBIOSYS
Certificates
undefined
Matrix Plus, CELLvo™
Supplier:  STEMBIOSYS

Specifications

  • Source:
  • Species:
  • Environmentally Preferable:

Specifications

Frequently Bought Together