Toluene ≥99.8% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 4483-25
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (C₆H₅CH₃ by GC):≥99.8 %
- Water (H₂O by KF, coulometric):≤0.01 %
- Titrable Acid (meq/g):≤0.0005 %
- Residue after Evaporation:≤0.0005 %
- Absorbance - 290 nm:≤0,3
- Absorbance - 300 nm:≤0,1
- Absorbance - 310 nm:≤0.05
- Absorbance - 330 nm:≤0.01
- Absorbance - 350 nm:≤0.004
Specification Test Results
About this item
:
:92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
:GHS02
:GHS08
:GHS07
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512