ContactUsLinkComponent
VWR®, Dispersing Aggregates
VWR®, Dispersing Aggregates
Supplier:  VWR Collection
Certificates
undefined
VWR®, Dispersing Aggregates
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 431-0289
  • 431-0281
  • 431-0280
  • 431-0282
  • 431-0283
  • 431-0284
  • 431-0285
  • 431-0286
  • 431-0278
  • 431-0287
  • 431-0279
  • 431-0288
  • Suitable for dispersing instrument:
  • Homogeniser M & L
  • Homogeniser S & M
  • Homogeniser M & L
  • Homogeniser M & L
  • Homogeniser M & L
  • Homogeniser XL
  • Homogeniser XL
  • Homogeniser S
  • Homogeniser S
  • Working range:
  • 50 - 2000 ml
  • 5 - 250 ml
  • 100 - 4000 ml
  • 300 - 7000 ml
  • 500 - 10000 ml
  • 2000 - 20000 ml
  • 5000 - 20000 ml
  • 0,1 - 5 ml
  • 0,3 - 10 ml