Specifikace
- Serum format:Frozen
- Description:Characterised FBS, US Origin, human mesenchymal stem cell screened
- Origin:US
- Haemoglobin:≤25 mg/dl
- Endotoxin:≤25 EU/ml
- Storage temperature:Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Pk:500 ml
Specifikace
O tomto produktu
Enhance human mesenchymal stem cell growth.
- Designed for human mesenchymal stem cells
- Sourced in the USA with complete traceability back to original source
- Low in antibodies and high in growth factors
- Virus panel testing according to 9 CFR 113,53
- 40 nm filtration
Each lot is screened by growing human mesenchymal stem cells (hMSC) over a minimum of three subculture generations with 10 percent serum. Human MSC are then observed for evidence of nutritional deficiency, cytotoxicity or morphological aberrations. The serum is tested in parallel with a control lot which has been demonstrated to support the growth of hMSC.
Data shows that 40 nm filtration will remove as many as eight logs of viral challenge. Studies demonstrate that this filtration regimen has no adverse effect on cell growth. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles. Defined FBS has over 50 components analyzed on the finished product and the results are included in the certificate of analysis and the biochemical assay list.