Order Entry
Czech Republic
ContactUsLinkComponent
 
Embryonic stem cell screened fetal bovine serum (FBS), U.S. origin, Hyclone™
 
undefined
Embryonic stem cell screened fetal bovine serum (FBS), U.S. origin, Hyclone™

 

  • Catalog No:
  • SH30071.03E
  • SH30071.02E
  • Origin:
  • US
  • US
  • Endotoxin:
  • ≤25 EU/ml
  • ≤25 EU/ml
  • Haemoglobin:
  • ≤25 mg/dl
  • ≤25 mg/dl
  • Storage temperature:
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.

 

Frequently Bought Together