ContactUsLinkComponent
Cleanroom gloves, nitrile, non sterile, white, smooth finish, PUREZERO® HG3
 :  Halyard
undefined
Cleanroom gloves, nitrile, non sterile, white, smooth finish, PUREZERO® HG3
 :  Halyard
  • Catalog No:
  • 112-6522
  • 112-6519
  • 112-6520
  • 112-6523
  • 112-6521
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Smooth, with textured fingertips
  • Smooth, with textured fingertips
  • Smooth, with textured fingertips
  • Smooth, with textured fingertips
  • Smooth, with textured fingertips
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Finger thickness:
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • Palm thickness:
  • 0,13 mm
  • 0,13 mm
  • 0,13 mm
  • 0,13 mm
  • 0,13 mm
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +
Frequently Bought Together