ContactUsLinkComponent
CrossLab Bottle Head Assemblies for HPLC
CrossLab Bottle Head Assemblies for HPLC
  HEWL01018-60025
 :  Agilent
undefined
CrossLab Bottle Head Assemblies for HPLC
  HEWL01018-60025
 :  Agilent
  • Description:
    Solvent inlet filter, stainless steel
  • Seq:
    24
  • Pk:
    1
All the parts required to connect solvent bottles to your HPLC instrument.
  • Instrument specific ready-to-use bottle head assemblies available
  • Special ultraclean tubing for MS applications available
Agilent bottle head assemblies contain all the parts that are required to connect solvent bottles to your HPLC instrument. Each assembly contains a solvent inlet filter, tubing, a bottle cap, and fittings.