Zadání objednávky
Czech Republic
ContactUsLinkComponent
PCR expansion
  OXNTEXP-PCA001
 :  
undefined
PCR expansion
  OXNTEXP-PCA001
 :  
  • Bal.:
    1 KIT
  • Popis:
    PCR expansion
  • Kompatibilní s:
    R10.4.1 flow cells
  • No. of reactions:
    24
PCR Primers (PRM) and PCR Adapters (PCA).
  • Additional PCR primers and adapters for use with Ligation Sequencing Kits
This kit contains PCR Primers and PCR Adapters, sufficient for at least 24 reactions, and is compatible for use with the Ligation Sequencing Kits (OXNTSQK-LSK110 and OXNTSQK-LSK109).
 : Flow cells and kits are shipped together at 2 to 8 °C. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.
 : Oxford Nanopore Technologies products are currently for research use only.