Sodium hexametaphosphate, Biotech Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:10124-56-8
Formula:Na₆P₆O₁₈
MDL Number:MFCD00136045
:611.77 g/mol
Specification Test Results
- Identification:Passes Test
- Appearance (white to off-white solid):Passes Test
- pH of 1% Solution at 25°C:6.7 - 7.2
- Loss on Drying at 105°C:≤ 0.5 %
- Assay (as P₂O₅):60.0 - 71.0 %
- Total Aerobic Microbial Count:≤ 100 cfu/g
- Total Yeasts and Molds:≤ 100 cfu/g
- Residual Methanol:≤ 3000 ppm
- Residual Ethanol:≤ 5000 ppm
- Residual Isopropanol:≤ 5000 ppm
Specification Test Results
Frequently Bought Together