Order Entry
Czech Republic
ContactUsLinkComponent
Benchtop cooler, StrataCooler LP
Benchtop cooler, StrataCooler LP
Catalog # AGLS401349
Supplier:  AGILENT
undefined
Benchtop cooler, StrataCooler LP
Catalog # AGLS401349
Supplier:  AGILENT

Specifications

  • Capacity:
    32×1,5 ml
  • Description:
    StrataCooler LP benchtop cooler, blue
  • W×D×H:
    187×120×80 mm
  • Pk:
    1

Specifications

About this item

The StrataCooler LP benchtop cooler provides the most protection for enzymes whether in freezer or on benchtop.

  • Maintains –15 °C for at least 2 hours
  • Eliminates ice chips and potential contamination

Cooler with slots for holding 1,5 ml and 0,5 ml tubes to keep temperature-sensitive enzymes cold while working at the bench.

Delivery: Cooler includes chilled lid, adapters and a locking handle.