Order Entry
Czech Republic
ContactUsLinkComponent
VWR®, Cell culture media, McCoy's 5A medium
VWR®, Cell culture media, McCoy's 5A medium
Catalog # 392-0420
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Cell culture media, McCoy's 5A medium
Catalog # 392-0420
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Pk:
    500 mL
  • Description:
    McCoy's 5A with L-Glutamine
  • Calcium:
    Yes
  • Magnesium:
    Yes
  • HEPES:
    No
  • Sodium bicarbonate:
    Yes
  • Sodium pyruvate:
    No
  • Phenol red:
    Yes
  • Stable glutamine:
    No
  • L-Glutamine:
    Yes
  • Media Base:
    McCoy's 5A Medium

Specifications

About this item

McCoy’s media were originally formulated for growth and support of lymphocytes. This final modification produced a medium identical to RPMI 1629. McCoy's 5A media supports the indefinite proliferation of Walker 256 carcinoma cells. In addition, it is excellent for the propagation of leukocytes, biopsy tissues, a broad range of human and rat normal or transformed cell types, the most current primary and continuous cell lines.