Zadání objednávky
Czech Republic
ContactUsLinkComponent
VWR®, Cell culture media, Ham's F12
VWR®, Cell culture media, Ham's F12
Katalogové číslo 392-0418
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Cell culture media, Ham's F12
Katalogové číslo 392-0418
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Formát média:
    Liquid
  • Bal.:
    500 mL
  • Popis:
    Ham's F12 with L-Glutamine
  • Vápník:
    Yes
  • Hořčík:
    Yes
  • HEPES:
    No
  • Hořčík:
    Yes
  • Hořčík:
    Yes
  • Hořčík:
    Yes
  • Stable glutamine:
    No
  • Hořčík:
    Yes
  • Media Base:
    Ham's F12 Nutrient Mixture
Specifications
About this item
Ham's F12 was developed for growth of primary rat hepatocytes and rat prostate epithelial cells. Also used in a clonal toxicity assay using CHO cells. It is frequently used with supplements for serum-free cultures to support the growth of cells of rodent origin, or as a cloning medium for the culture of myeloma and hybrid cells.