Peristaltic pump tubing and sample line supplies
Supplier: Agilent
Certificates
About this item
The peristaltic pump tubing has to be washed prior to use. The common way is to run about 5% HNO₃ (v/v) solution.
- Long lifetime
- Resistant to most chemicals
- Not suitable for sample introduction - impurity level is high
If customers use HCl, Sn is eluted from the Tygon tube. In this case, 3% HCl (v/v) should be pumped at 0,1 rps for less than an hour by the new peristaltic pump tube.
Caution: Some chemicals attack the peristaltic pump tube. This might cause leakage in the sample introduction and drain lines and corrosion of the sample introduction system.
Frequently Bought Together