Order Entry
Czech Republic
ContactUsLinkComponent
Etoposide 100 mM
Etoposide 100 mM
Catalog # API-06
Supplier:  G-Biosciences
CAS Number:  
undefined
Etoposide 100 mM
Catalog # API-06
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    50 µl
  • Seq:
    1202

Specifications

About this item

 :
 :588.56 g/mol
CAS Number:33419-42-0
Formula:C₂₉H₃₂O₁₃
MDL Number:MFCD00869325