ContactUsLinkComponent
Tools for PerkinElmer
  HEWL8003-0423
 :  Agilent
Tools for PerkinElmer
  HEWL8003-0423
 :  Agilent
  • Description:
    Insertion tool for L'vov platforms
  • Pk:
    1
These tools are guaranteed to be compatible with PerkinElmer instruments.
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Instruction guides and how-to videos available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
Keep Agilent tools in your toolbox to maximise uptime of your PerkinElmer systems.