Order Entry
Czech Republic
ContactUsLinkComponent
Single element standards for ICP-OES, magnesium
Single element standards for ICP-OES, magnesium
Catalog # HEWL5190-8412
Supplier:  Agilent
CAS Number:  
undefined
Single element standards for ICP-OES, magnesium
Catalog # HEWL5190-8412
Supplier:  Agilent
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    5% HNO₃
  • Pk:
    100 mL
  • Element:
    Magnesium
  • Symbol:
    Mg

Specifications

About this item

Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.

  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations

All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.

Delivery: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Packaging: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.