Order Entry
Czech Republic
ContactUsLinkComponent
Propylene carbonate 99%
Irritant
Propylene carbonate 99%
Catalog # ABCRAB119095.0250
Supplier:  abcr
CAS Number:  
Irritant
Propylene carbonate 99%
Catalog # ABCRAB119095.0250
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

CAS Number:108-32-7
 :
Formula:C₄H₆O₃
MDL Number:MFCD00005385
 :102.09 g/mol
 :GHS07
Storage Temperature:Ambient