ContactUsLinkComponent
Accessories for sample loops for Rheodyne® valves
Accessories for sample loops for Rheodyne® valves
Catalog # HEWL5068-0099
Supplier:  Agilent
Accessories for sample loops for Rheodyne® valves
Catalog # HEWL5068-0099
Supplier:  Agilent
Specifications
  • Description:
    Rotor, 12-position/13-port bio valve, 200 bar
  • Seq:
    02
  • Pk:
    1