ISIS supplies for 7700
Catalog # HEWLG4911-01201
Supplier: Agilent
Specifications
- For:Agilent 7700/8000 series ICP-MS
- Description:Joint holder for nebuliser pump
- Pk:1
Specifications
About this item
A range of high quality supplies and components for the Integrated Sample Introduction System (ISIS) for use with the 7700 ICP-MS range.
- A variety of joints are available in tee, cross or union form
Agilent's ISIS enhances the functionality and flexibility of the Agilent 7700 series ICP-MS. ISIS features high precision peristaltic pumps and an optional switching valve to allow complete flexibility of configuration. The ISIS hardware is built into the 7700 chassis and close-coupled to increase productivity through high-speed sample uptake and rapid washout. To enhance the capabilities that the system offers and maintain performance even after heavy use, Agilent provides a range of replacement components and other supplies.