Order Entry
Czech Republic
ContactUsLinkComponent
 
Single element standard for ICP-MS, Uranium
  HEWL5190-8584
 :  Agilent
 :  
Single element standard for ICP-MS, Uranium
  HEWL5190-8584
 :  Agilent
 :  

 

  • Concentration:
    10 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Element:
    Uranium
  • Symbol:
    U

 

 

Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionization buffers and environmental standards for Atomic Spectroscopy.

  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations

All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.

 : Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
 : All standards packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.