Vanadium, single element standard for ICP-MS
Catalog # HEWL5190-8585
Supplier: Agilent
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:5% HNO₃
- Pk:100 mL
- Element:Vanadium
- Symbol:V
Specifications
About this item
It offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for atomic spectroscopy.
- It is works in AAS, ICP-OES or ICP-MS and MP-AES results
- Accurate, reliable calibrations
Standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.