Order Entry
Czech Republic
ContactUsLinkComponent
GC columns, J&W VF-5ms
GC columns, J&W VF-5ms
Catalog # HEWLCP9043
Supplier:  Agilent
undefined
GC columns, J&W VF-5ms
Catalog # HEWLCP9043
Supplier:  Agilent

Specifications

  • Film thickness:
    0,25 µm
  • Ø int.:
    0,25 mm
  • Length:
    20 m
  • Phase:
    VF-5 ms
  • Pk:
    1

Specifications

About this item

VF-5 ms is an inert and low bleed 5% phenyl-methyl phase for increased sensitivity, accuracy, and instrument uptime for semipolar and polar compounds.

  • 0.15 mm int.Ø columns available for high efficiency GC and GC/MS analysis
  • QC test results for retention index, efficiency, selectivity and bleed is reported with every column

The column has the lowest guaranteed bleed specification of 1 pA at 325 °C (30 m, 0.25 mm, 0.25 µm). VF-5ms has a slightly higher polarity than the VF-1 ms, resulting in a better selectivity for aromatic compounds. This selectivity, combined with superior inertness, also makes these columns applicable for a wide range of semipolar and even polar components, such as phenols.