
alpha-Biotin-omega-carboxysuccinimidyl ester poly(ethyleneglycol), M.W. PEG molecular weight 5,000 daltons
Catalog # APOSBIP1057-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:GHS07
Storage Temperature:Freezer