Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Recombinant urate oxidase
Recombinant urate oxidase
Recombinant urate oxidase
Catalog # APOSBITP1363-1MG
Dodavatel:
Apollo Scientific
Číslo CAS:
Expand image
Recombinant urate oxidase
Catalog # APOSBITP1363-1MG
Dodavatel:
Apollo Scientific
Číslo CAS:
Specifikace
Pk:
1 mg
Enzyme type:
Recombinant
Storage conditions:
Ambient
Enzyme name:
Urate Oxidase
Enzyme synonyms:
UOX
Specifikace
O tomto produktu
Product Details
Product Details