Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Home
Antibodies
Anti-c-myc Mouse Antibody
Anti-c-myc Mouse Antibody
Anti-c-myc Mouse Antibody
Catalog # APOSBITP2048-1MG
Supplier:
Apollo Scientific
Rozbalit obrázek
Anti-c-myc Mouse Antibody
Catalog # APOSBITP2048-1MG
Supplier:
Apollo Scientific
Specifications
Antigen symbol:
c-myc
Host:
Mouse
Reactivity:
Human
Pk:
1 mg
Specifications
About this item
Product Details
Product Details