Order Entry
Czech Republic
ContactUsLinkComponent
Anti-NT-4 Mouse Antibody
Anti-NT-4 Mouse Antibody
Catalog # APOSBITP1986-1MG
Supplier:  Apollo Scientific
Anti-NT-4 Mouse Antibody
Catalog # APOSBITP1986-1MG
Supplier:  Apollo Scientific

Specifications

  • Antigen name:
    Neurotrophin-4
  • Antigen symbol:
    NT-4
  • Host:
    Mouse
  • Reactivity:
    Human
  • Pk:
    1 mg

Specifications

About this item