Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Poly(caprolactone-co-glycolide)
Poly(caprolactone-co-glycolide) 45:55
Poly(caprolactone-co-glycolide) 45:55
Poly(caprolactone-co-glycolide) 45:55
Catalog # APOSBIBE1004-5G
Supplier:
Apollo Scientific
CAS Number:
Expand image
Poly(caprolactone-co-glycolide) 45:55
Catalog # APOSBIBE1004-5G
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pack type:
Bottle
Pk:
5 g
Seq:
3173
Specifications
About this item
Product Details
:
Product Details