Order Entry
Czech Republic
ContactUsLinkComponent
 
Human recombinant PCNA
Catalog # APOSBITP2123-5UG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant PCNA
Catalog # APOSBITP2123-5UG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    5 g
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    03
  • Protein synonyms:
    Proliferating Cell Antigen
  • Protein/peptide name:
    PCNA

Specifications

About this item