Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Rainbow traut recombinant Prl-R
Rainbow traut recombinant Prl-R
APOSBITP2096-1MG
:
Apollo Scientific
:
Rainbow traut recombinant Prl-R
APOSBITP2096-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Rainbow trout
Protein synonyms:
Raibow Trout Prolactin Soluble Receptor
Protein/peptide name:
Prl-R