Recombinant HIV-2 gp32, HRP (Horseradish Peroxidase)
Supplier: Apollo Scientific
Certificates
About this item
Specifications
- Conjugation:HRP (Horseradish Peroxidase)
- Protein/peptide type:Recombinant
- Protein/peptide name:HIV-2 gp32
Specifications
Frequently Bought Together