0,00 Kč
Zadání objednávky
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Proteiny
Recombinant HIV-1 pol Integrase
Recombinant HIV-1 pol Integrase
Catalog # APOSBITP1891-100UG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Recombinant HIV-1 pol Integrase
Catalog # APOSBITP1891-100UG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Bal.:
100 µG
Typ proteinu/peptidu:
Recombinant
Proteinová synonyma:
HIV-1 Integrase
Název proteinu/peptidu:
HIV-1 pol Integrase
Specifications
About this item
Product Details
Product Details