Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant HIV-I p24 gag
Recombinant HIV-I p24 gag
APOSBITP1872-100UG
:
Apollo Scientific
:
Recombinant HIV-I p24 gag
APOSBITP1872-100UG
:
Apollo Scientific
:
Pk:
100 µG
Protein/peptide type:
Recombinant
Protein synonyms:
HIV-1 p24 gag
Protein/peptide name:
HIV-I p24 gag