Human recombinant Fragile Histidine Triad
Catalog # APOSBITP1558-5UG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:5 µG
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:FHIT
- Protein/peptide name:Fragile Histidine Triad