0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Proteins and Peptides
Human recombinant C-reactive protein
Human recombinant C-reactive protein
APOSBITP1529-1MG
:
Apollo Scientific
:
Human recombinant C-reactive protein
APOSBITP1529-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Human
Seq:
23
Protein synonyms:
CRP
Protein/peptide name:
C-Reactive Protein