Human Recombinant ERK2/MAPK1 Inactive Enzyme
Catalog # APOSBITP1402-10UG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:ERK2/MAPK1
- Protein/peptide name:ERK2/MAPK1 Inactive Enzyme