Human Recombinant Casein Kinase 2 Holoenzyme
Catalog # APOSBITP1399-20UG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:CK2h
- Protein/peptide name:Casein Kinase 2 Holoenzyme