Order Entry
Czech Republic
ContactUsLinkComponent
Human Recombinant Epidermal Growth Factor Receptor (from Sf9 cells)
Human Recombinant Epidermal Growth Factor Receptor (from Sf9 cells)
Supplier:  Apollo Scientific
Certificates
Human Recombinant Epidermal Growth Factor Receptor (from Sf9 cells)
Supplier:  Apollo Scientific

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Human
  • Protein synonyms:
    EGFR
  • Protein/peptide name:
    Epidermal Growth Factor Receptor

Specifications

Frequently Bought Together