0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Proteins and Peptides
Sheep Recombinant Prl
Sheep Recombinant Prl
Catalog # APOSBITP1248-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Sheep Recombinant Prl
Catalog # APOSBITP1248-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Sheep
Protein synonyms:
Prolactin
Protein/peptide name:
Prl
Specifications
About this item
Product Details
Product Details