Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Sheep Recombinant PL
Sheep Recombinant PL
Sheep Recombinant PL
APOSBITP1241-1MG
:
Apollo Scientific
:
Sheep Recombinant PL
APOSBITP1241-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Sheep
Protein synonyms:
Placental Lactogen
Protein/peptide name:
PL