Human Recombinant PDGF-BB (from Yeast)
Catalog # APOSBITP1234-10UG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:Yeast
- Species:Human
- Seq:05
- Protein synonyms:Platelet Derived Growth Factor-BB
- Protein/peptide name:PDGF-BB